Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (Cy3®)

Supplier: Cytiva
28-9011-06 28-9011-06
95040-030EA 621.52 USD InStock
CA95040-030L 95040-030
Anti-IgG Goat Polyclonal Antibody (Cy3®)
Antibodies
ECL Plex Cy-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent western blotting.

  • For 1000 cm² membrane area

Optimized system for quantitative analysis using fluorescent Western blotting. ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.

ECL Plex Western Blotting System is optimized for single protein detection as well as multiplex protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.

Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty