Would you like to log in again?
Mouse Recombinant IL31 (from E. coli)
Supplier: BioVision
AB285697-1MG AB285697-100UG AB285697-10UG
10007-372EA 757.27 USD InStock
10007-372 10007-374 10007-376
Mouse Recombinant IL31 (from E. coli)
Proteins and Peptides
>97% Pure Recombinant murine IL-31.
A standard activity has not yet been determined.
Interleukin-31 (IL-31) is an immunoregulatory cytokine that is mainly expressed by activated TH2 cells. IL-31 signals through the IL-31 Receptor (IL-31RA) and the oncostain receptor (OSMR), which are expressed on monocytes, epithelial cells, and keratinocytes. IL-31 has a role in inflammatory skin diseases. Recombinant mouse IL-31 is a non-glycosylated protein, containing 141 amino acids, with a mass of 15.7 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty