Would you like to log in again?
Human Recombinant Galectin 3 (from E. coli)
Supplier: BioVision
AB285623-1MG AB285623-10UG AB285623-50UG
10006-980EA 207.23 USD InStock
10006-980 10006-984 10006-982
Human Recombinant Galectin 3 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human Galectin-3.
Determined by its ability to chemoattract human blood monocytes. Chemotactic activity was observed at a concentration of 2.5 µg/ml with a peak response obtained at 250 µg/ml.
Lectins are carbohydrate binding proteins that interact with glycoprotein and glycolipids on the surface of animal cells. The Galectins are lectins that recognize and interact with β-galactoside moieties. Galactin-3 regulates a number of biological processes, including embryogenesis, inflammatory responses, cell progression and metastasis. Galectin-3 can function intraCellularly in controlling cell cycle and preventing T-cell apoptosis, and also extraCellularly, in activating various cells, including monocytes/macrophages, mast cells, neutrophils, and lymphocytes. Recombinant Galectin-3 is a globular 26.0 kDa protein containing 250 amino acid residues, but no disulfide bonds.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty