Would you like to log in again?
Human Recombinant Dipeptidylpeptidase Iv (from Hi-5 Insect Cells)
Supplier: BioVision
AB129472-10UG AB285644-1MG AB129472-50UG
10007-016EA 397.75 USD InStock
10007-016 10007-018 10007-020
Human Recombinant Dipeptidylpeptidase Iv (from Hi-5 Insect Cells)
Enzymes
>95% Pure Recombinant human Dipeptidylpeptidase IV.
>20 Units/mg
DPP4 is a serine exopeptidase and complex enzyme that is expressed on the surface of most cell types as an intrinsic membrane glycoprotein that cleaves X-proline dipeptides from the N-terminus of polypeptides and plays a role in t-cell activation. DPP4 is associated with intracellular signal transduction, apoptosis and involved in tumor biology. There are at least 63 substrates which can bind specifically to DPP4 enzyme including growth factors, chemokines, neuro peptides. Furthermore, DPP4 plays a major role in glucose metabolism by cleaving incretins such as glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1). DPP4 Human Recombinant is a single, glycosylated polypeptide chain containing 746 amino acids (aa 39-766) with a C-terminal His-tag and is purified using conventional chromatography techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty