Would you like to log in again?
Human Recombinant Growth hormone (from E. coli)
Supplier: BioVision
AB285654-500UG AB285654-100UG AB285654-5MG AB285654-1MG
10007-126EA 542.51 USD InStock
10007-126 10007-128 10007-130 10007-124
Human Recombinant Growth hormone (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human Growth Hormone.
BioVision's pituitary Growth Hormone is fully biologically active when compared to World Health Organization (WHO) reference standard.
Recombinant Human Pituitary Growth Hormone produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 192 amino acids and having a molecular mass of 22367 Dalton. Pituitary Growth Hormone is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty