Would you like to log in again?
Human Recombinant Ghrh (from E. coli)
Supplier: BioVision
AB52521-500
10007-152EA 347.56 USD InStock
10007-152
Human Recombinant Ghrh (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human FSH.
BioVision's hGHRH is fully biologically active, increases plasma growth hormone concentrations by directly stimulating the anterior pituitary gland to release natural human growth hormone.
The recombinant human GHRH is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular mass of 3358 Dalton. Corresponds to the amino-terminal segment of the naturally occurring human growth hormone-releasing hormone consisting of 44 amino acid residues. rhGHRH is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty