Would you like to log in again?
Human Recombinant Superoxide Dismutase (from E. coli)
Supplier: BioVision
AB112193-100UG
10007-168EA 741.99 USD InStock
10007-168
Human Recombinant Superoxide Dismutase (from E. coli)
Enzymes
>98% Pure active Recombinant human Superoxide Dismutase.
BioVision recombinant Superoxide Dismutase is fully biologically active when compared to standard. Activity tests were carried using SOD activity assay kit (K335-100). The activity assay kit showed that the calculated activity was ~40,000 U/mg. This was calculated from the amount (µg) required to inhibit the Xanthine Oxidase activity by 50%
Superoxide Dismutase (SOD) is an oxidoreductase that catalyzes the reaction between superoxide anions and hydrogen to yield molecular oxygen and hydrogen peroxide. The enzyme protects the cell against dangerous levels of superoxide. Recombinant Human Cu/Zn Superoxide Dismutase produced in E.Coli is a homodimer, non-glycosylated polypeptide chain containing 2 x 154 amino acids and having a total molecular mass of 31,608 Dalton. The Cu/Zn SOD is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty