Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sirtuin Activity Assay Kit Fluorometric, BioVision

Supplier: BioVision
AB287850-96TEST
10013-678EA 842.96 USD InStock
10013-678
Sirtuin Activity Assay Kit Fluorometric, BioVision
Assays Protein Assays
Validated using many sample types

  • Detection method- Fluorescence (Ex/Em = 400/505 nm)
  • Application- Detection of Sirtuin Activity in variety of samples

Sirtuins are a class of proteins that possess either histone deacetylase or mono-ribosyltransferase activity. Sirtuins are localized in the cytoplasm, nucleus, nucleolus as well as mitochondria. They are associated with aging, cellular protection, sugar metabolism and cell cycle regulation. Unlike other known protein deacetylases, which simply hydrolyze acetyl-lysine residues, the sirtuin-mediated deacetylation reaction hydrolyzes acetyl-lysine and NAD. This hydrolysis yields the deacetylated substrate, O-acetyl-ADP-ribose and nicotinamide, itself an inhibitor of sirtuin activity. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. In BioVision’s Sirtuin Activity Assay Kit, the acetylated p53-AFC substrate is deacetylated by Sirtuins in the presence of NAD+ to generate the deacetylated p53-AFC substrate, nicotinamide and O-Acetyl-ADP Ribose. Cleavage of the deacetylated p53-AFC substrate by the Developer releases the fluorescent group, which is detected fluorometrically at Ex/Em = 400/505 nm. HDAC’s also deacetylate the acetylated p53-AFC substrate. Trichostatin A is added to the reaction to specifically inhibit HDAC’s in samples. This kit provides a rapid, simple, sensitive, and reliable test to measure Sirtuin Activity in a variety of samples. The limit of quantification of the assay is 0.06 µU of recombinant human SIRT6.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty