Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urease Activity Assay Kit Colorimetric, BioVision

Supplier: BioVision
AB204697-100TEST
10013-694EA 892.99 USD InStock
10013-694
Urease Activity Assay Kit Colorimetric, BioVision
Assays Protein Assays
Validated using yeast, bacteria and soil samples

  • Detection method- Absorbance (670 nm)
  • Application:
  • - Measurement of urease activity

Urease (EC 3.5.1.5) catalyzes the hydrolysis of urea into carbon dioxide and ammonia. Urease is found in certain bacteria, yeast, fungi, and plants. In addition, it is found environmentally in soil samples and fecal matter. Due to ammonia production, urease can play a role in the pathogenesis of certain medical conditions including urinary stones, peptic ulcers, and hepatic encephalopathy. As such, urease can be used as a diagnostic for gastrointestinal and urinary tract pathogens. BioVision’s Urease Activity Assay Kit provides a quick and easy way to determine urease activity. The kit uses a modified Berthelot method to detect ammonia, which can be measured at 670 nm. The kit is suitable for measuring urease activity of biological and soil samples as well as of purified enzyme. The limit of detection is below 0.001 mU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty