Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plasmin Activity Assay Kit Fluorometric, BioVision

Supplier: BioVision
AB204728-100TEST
10013-696EA 741.98 USD InStock
10013-696
Plasmin Activity Assay Kit Fluorometric, BioVision
Assays Protein Assays
Validated using plasma samples

  • Detection method- Fluorescence (Ex/Em = 360/450 nm)
  • Applications- Determine activity of pure Plasmin
  • - Detect the activity of Plasmin in plasma

Plasmin (EC 3.4.21.7) is a serine protease occurring in plasma as plasminogen. Upon activation via cleavage by plasminogen activators; plasmin solubilizes fibrin clots and activates and/or degrades compounds of the coagulation and complement systems. Plasminogen-Plasmin system has also been implicated in a wide variety of physiologic and pathologic processes, including tumor growth, invasion and metastasis. BioVision's Plasmin activity assay kit utilizes the ability of Plasmin to proteolytically cleave a synthetic substrate and release a fluorophore, AMC, which can be easily quantified by fluorescence microplate readers. This assay kit is simple, rapid and can detect Plasmin activity as low as 10 ng in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty