Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plasmin Inhibitor Scree Kit Fluorometric, BioVision

Supplier: BioVision
AB204729-100TEST
10013-698EA 980.59 USD InStock
10013-698
Plasmin Inhibitor Scree Kit Fluorometric, BioVision
Assays Protein Assays
Validated using aprotinin inhibitor

  • Detection Method – Fluorescence (Ex/Em = 350/450 nm)
  • Application:
  • - Screening potential inhibitors of plasmin
  • - Characterize/study plasmin inhibitors

Plasmin (EC 3.4.21.7) is a serine protease occurring in plasma as plasminogen. Upon activation via cleavage by plasminogen activators; plasmin solubilizes fibrin clots and activates and/or degrades compounds of the coagulation and complement systems. Plasmin inhibitors are critical in the treatment of hyperfibrinolysis-associated blood loss and related complications. Biovision’s Plasmin Inhibitor Screening Kit utilizes the ability of Plasmin to cleave a synthetic AMC-based peptide substrate to release AMC, which can be detected by measuring its fluorescence at Ex/Em = 360/450 nm. In the presence of Plasmin specific inhibitors, the extent of cleavage reaction is reduced or completely abolished. The loss in the fluorescence intensity can be correlated to the amount of inhibitor present in the assay solution. The kit provides a simple and rapid method to screen potential inhibitors of Plasmin.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty