Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neutrophil Elastase Activity Assay kit (Fluorometric), BioVision

Supplier: BioVision
AB204730-100TEST
10013-700EA 684.71 USD InStock
10013-700
Neutrophil Elastase Activity Assay kit (Fluorometric), BioVision
Assays Protein Assays
Validated using blood samples

  • Detection method- Fluorescence (Ex/Em = 380/500 nm)
  • Applications- Determine activity of purified Neutrophil Elastase
  • - Detect the activity of Neutrophil Elastase in plasma, blood
  • - Screen/study/characterize chemotactic agents causing Neutrophil stimulation and release of Elastase

Neutrophil Elastase (NE, EC 3.4.21.37, leukocyte elastase, elastase 2, or serine elastase) is a cytotoxic serine protease. It is stored in the azurophil granules of neutrophil granulocyte and is released following cell stimulation, e.g. by pathogens, immune complexes or chemotactic agents (PMA). When the extracellular NE concentration exceeds the buffering capacity of endogenous inhibitors, it causes degradation of a wide range of extracellular matrix proteins, including fibronectin, laminin, proteoglycans, collagens, and elastin. Extracellular elastase is implicated in the signs, symptoms and disease progression of inflammatory lung disorders (such as cystic fibrosis, COPD, lung emphysema) via its role in the inflammatory processes, mucus overproduction and lung tissue damage. BioVision's Neutrophil Elastase Activity Assay Kit utilizes the ability of NE to proteolytically cleave a synthetic substrate and release a fluorophore, AFC, which can be easily quantified by fluorescence microplate readers. This assay kit is simple, rapid and can detect as low as 1 ng of Neutrophil Elastase in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty