Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfatase Activity Assay Kit Colorimetric, BioVision

Supplier: BioVision
AB204731-100TEST
10013-770EA 1174.83 USD InStock
10013-770
Sulfatase Activity Assay Kit Colorimetric, BioVision
Assays Protein Assays
Only kit on the market to measure sulfatase activity

  • Detection method- Absorbance (515 nm)
  • Application - Measurement of sulfatase activity

Sulfatases (EC 3.1.6) are enzymes in the esterase class that catalyze the hydrolysis of sulfate esters from a wide range of biological molecules, including steroids, carbohydrates, and proteins. They can be found in intracellular and extracellular spaces and are distributed in a wide range of cells and tissues. Intracellular sulfatases are commonly found localized within the lysosome. Genetic defects in sulfatase can result in certain lysosomal storage disorders and abnormal expression can contribute to certain hormone-dependent cancers, such as breast and prostate cancer. BioVision’s Sulfatase Activity Assay Kit provides a quick and easy way to determine sulfatase activity. The kit measures the hydrolysis of a sulfate ester to 4-nitrocatechol, which can be detected at 515 nm. The kit is suitable for measuring activity of purified enzyme as well as sulfatase from biological samples. The limit of detection is below 1 mU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty