Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gapdh Activity Assay Kit, BioVision

Supplier: BioVision
AB204732-100TEST
10013-772EA 386.06 USD InStock
10013-772
Gapdh Activity Assay Kit, BioVision
Assays Protein Assays
Sensitive Assay, HTS

  • Detection method- Absorbance (450 nm)
  • Applications- - Measurement of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) activity in various tissues/cells
  • - Analysis of glycolysis and pentose phosphate pathways

GAPDH (Glyceraldehyde-3-Phosphate Dehydrogenase; EC 1.2.1.12) catalyzes the conversion of Glyceraldehyde-3-Phosphate (GAP) to 1, 3-Bisphosphate Glycerate (BPG) and plays a key role in glycolysis. The enzyme is involved in cellular processes such as apoptosis, membrane trafficking, iron metabolism and nuclear translocation. GAPDH (housekeeping gene) expression is stable and constitutive. Deregulation of GAPDH activity is associated with abnormal cell proliferation and carcinogenesis. Accurate quantitation of GAPDH activity is important for diagnosing diseases and studying normal cellular physiology. BioVision’s GAPDH Activity Assay Kit provides a simple and sensitive method for monitoring GAPDH activity in various samples. In this assay, GAPDH catalyzes conversion of GAP into BPG and an intermediate, which reacts with a developer to form a colored product that absorbs maximally at 450 nm. Our high-throughput adaptable assay can detect GAPDH activity as low as 100 µU/mL in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty