Would you like to log in again?
Human Recombinant CRP (from E. coli)
Supplier: BioVision
AB285671-1MG AB285671-250
10007-236EA 254.2 USD InStock
10007-236 10007-234
Human Recombinant CRP (from E. coli)
Proteins and Peptides
>95% Pure Recombinant human CRP.
Recombinant Human C-Reactive Protein produced in E. coli is a non-glycosylated polypeptide chain having a total molecular mass of 115kDa, corresponding to the pentamer structure of 23 kDa monomers determined by amino acid sequence. Recombinant Human CRP is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty