Would you like to log in again?
Annexin V-Egfp Apoptosis Kit, BioVision
Supplier: BioVision
AB14153-100
10013-270EA 1138.15 USD InStock
10013-270
Annexin V-Egfp Apoptosis Kit, BioVision
Assays Protein Assays
Detects Apoptosis in live cells within 10 min.
  • Flow cytometry (Ex = 488 nm; Em = 530 nm) and fluorescence microscopy
  • Mammalian
  • 25, 100, 400
  • Applications-Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis
The Annexin V-EGFP Apoptosis Detection Kit is based on the observation that soon after initiating apoptosis, most cell types translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can be easily detected by staining with an enhanced green fluorescent protein (EGFP) fusion of annexin V, a protein that has a strong natural affinity for PS. The one-step staining procedure takes only 10 minutes. In addition, the assay can be directly performed on live cells. Detection can be analyzed by flow cytometry or by fluorescence microscopy with a FITC filter. EGFP is brighter and more photo-stable than other fluorescent reagents. The kit can differentiate apoptosis vs necrosis when performing both annexin V-EGFP and PI staining.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty