Would you like to log in again?
Caspase-6 Colorimetric Assay Kit, BioVision
Supplier: BioVision
K115-200 AB39709-100 K115-400 K115-25
10013-536EA 974.85 USD InStock
10013-536 10013-534 10013-532 10013-530
Caspase-6 Colorimetric Assay Kit, BioVision
Assays Protein Assays
Convenient, Highly-Sensitive Assay
  • Absorbance (400 or 405 nm)
  • Mammalian
  • Convenient sizes (100 assays)
  • Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis
Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. The Caspase-6 Colorimetric Assay Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence VEID. The assay is based on spectrophotometric detection of the chromophore p-nitroanilide (pNA) after cleavage from the labeled substrate VEID-pNA. The pNA light emission can be quantified using a spectrophotometer or a microtiter plate reader at 400- or 405 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty