Would you like to log in again?
Human Recombinant Hexokinase 1 (from E. coli)
Supplier: BioVision
AB285719-100UG
10008-156EA 1800.29 USD InStock
10008-156
Human Recombinant Hexokinase 1 (from E. coli)
Enzymes
>90% Pure Recombinant Active Human Hexokinase 1
Specific activity is 7-8 units/ml obtained by measuring the increase of NADPH in absorbance at 340 nm resulting from the reduction of NADP.
Hexokinase is the first enzyme in the glycolytic pathway, catalyzing the transfer of a phosphoryl group from ATP to glucose to form glucose-6-phosphate and ADP. In vertebrates there are four major glucose-phosphorylating isoenzymes, designated hexokinase I, II, III, and IV This enzyme is found in most cells, but there is tissue specificity for the particular type of hexokinase. Hexokinase1 is found in the adipose tissue and liver and encodes a ubiquitous form of hexokinase which localizes to the outer membrane of mitochondria. Mutations in this hexokinase1 have been associated with hemolytic anemia due to hexokinase deficiency.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty