Would you like to log in again?
Caspase-9 Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB65607-100
10013-546EA 504.79 USD InStock
10013-546
Caspase-9 Fluorometric Assay Kit, BioVision
Assays Protein Assays
Convenient, Highly-Sensitive Assay
  • fluorecence (400 nm excitation filter and 505 nm emission filter)
  • Mammalian
  • Convenient sizes (25, 100, 200, 400 assays)
  • Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis
Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. The Caspase-9 Fluorometric Assay Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence LEHD. The assay is based on detection of cleavage of substrate LEHD-AFC (AFC: 7-amino-4-trifluoromethyl coumarin). LEHD-AFC emits blue light (λmax = 400 nm); upon cleavage of the substrate by caspase-9 or related caspases, free AFC emits a yellow-green fluorescence (λmax = 505 nm), which can be quantified using a fluorometer or fluorecence microtiter plate reader.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty