Would you like to log in again?
Human Recombinant PRDX2 (from E. coli)
AB167977-100UG
10008-176EA 888.69 USD InStock
10008-176
Human Recombinant PRDX2 (from E. coli)
Proteins and Peptides
>90% Pure Recombinant Active Human PRDX 2
Specific activity: approximately 200-230 pmole/min/µg.
Peroxiredoxin-2 is a protein that in humans is encoded by the PRDX2 gene. This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8 (+) T-cells. It may also have a proliferative effect and play a role in cancer development or progression.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty