Would you like to log in again?
Human Recombinant ALDH2 (from E. coli)
Supplier: BioVision
AB167991-100UG
10008-016EA 689.28 USD InStock
10008-016
Human Recombinant ALDH2 (from E. coli)
Proteins and Peptides
>90% Pure Recombinant Active Human ALDH2
Specific activity is > 0.14 units/ml
ALDH2 (Aldehyde dehydrogenase 2 family) belongs to the aldehyde dehydrogenase family which catalyze the chemical transformation from acetaldehyde to acetic acid and is the second enzyme of the major oxidative pathway of alcohol metabolism. There are two major liver isoforms of this enzyme, cytosolic and mitochondrial, and they can be also distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. ALDH2 has been shown to interact with GroEL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty