Would you like to log in again?
Human Recombinant ENO1 (from E. coli)
Supplier: BioVision
AB167976-100UG
10008-418EA 781.41 USD InStock
10008-418
Human Recombinant ENO1 (from E. coli)
Enzymes
  • Accession number: P06733
  • Gene ID: 2023
Alpha-enolase, also known as Enolase 1, is one of three enolase isoenzymes and a glycolytic enzyme expressed in most tissues. This protein plays a key role in anaerobic metabolism under hypoxic conditions and may act as a cell surface plasminogen receptor during tissue invasion. Abnormal expression of alpha-enolase is associated with tumor progression in some cases of breast and lung cancer. It also has been identified as an auto antigen associated with Hashimoto's encephalopathy and severe asthma.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty