Would you like to log in again?
Human Recombinant AKR1C3 (from E. coli)
Supplier: BioVision
AB167984-100UG
10008-366EA 866.38 USD InStock
10008-366
Human Recombinant AKR1C3 (from E. coli)
Proteins and Peptides
>98% Pure Recombinant Active Human AKR1C3
The specific activity of AKR1C3 with 5 mM 1-Indanol is approximately 0.3 U/mg.
Aldo-keto reductase family 1 member C3 in humans is encoded by the AKR1C3 gene. This enzyme catalyzes the reduction of prostaglandin (PG) D2, PGH2 and phenanthrenequinone (PQ), and the oxidation of 9alpha, 11beta-PGF2 to PGD2. It may play an important role in the pathogenesis of allergic diseases such as asthma, and may also have a role in controlling cell growth and/or differentiation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty