Would you like to log in again?
Human Recombinant Acid Phosphatase 1 (from E. coli)
Supplier: BioVision
AB168008-100UG
10008-446EA 1003.56 USD InStock
10008-446
Human Recombinant Acid Phosphatase 1 (from E. coli)
Enzymes
>95% Pure Active Recombinant Human Acid phosphatase 1
> 60,000 unit/mg of protein.
Acid phosphatase 1, also known as ACP1, belongs to the phosphotyrosine protein family. It functions as an acid phosphatase and a protein tyrosine phosphatase (PTPase) and is present in all human tissues, including adipocytes. This enzyme hydrolyzes protein tyrosine phosphate to protein tyrosine and orthophosphate, and also orthophosphoric monoesters to alcohol and orthophosphate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty