Would you like to log in again?
Mouse Recombinant Dhfr (from E. coli)
Supplier: BioVision
AB168723-100UG
10008-488EA 796.72 USD InStock
10008-488
Mouse Recombinant Dhfr (from E. coli)
Proteins and Peptides
>95% Pure Active Recombinant Mouse DHFR
Specific activity is > 0.2 units/mg and was obtained by measuring the oxidation of NADPH in absorbance at 340 nm during reaction.
Dihydrofolate reductase (DHFR) is a member of the reductase family of enzymes that is ubiquitously expressed in all organisms. DHFR catalyzes the NADPH-dependent reduction of dihydrofolate to tetrahydrofolate, and it is essential for the synthesis of thymidylate, purines and several amino acids. Expression of methotrexate (MTX)-resistant variants of DHFR in normal hematopoietic cells is a potential strategy to permit administration of larger doses of MTX by alleviating drug toxicity in normal cells and tissues that are drug sensitive.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty