Would you like to log in again?
Human Recombinant AK1 (from E. coli)
Supplier: BioVision
AB167983-100UG
10008-492EA 796.73 USD InStock
10008-492
Human Recombinant AK1 (from E. coli)
Proteins and Peptides
>95% Pure Active Recombinant Human AK1
Specific activity: > 6.0 units/ml.
AK1 is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of the terminal phosphate group between ATP and AMP. This protein is found in the cytosol of skeletal muscle, brain and erythrocytes. It is a small ubiquitous enzyme which is essential for maintenance and cell growth. Defects in AK1 are the cause of a form of hemolytic anemia. Recombinant human AK1 protein, fused to His-tag at N-terminus, was expressed in E. coli and purified by using conventional chromatography.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty