Would you like to log in again?
Human Recombinant Glyoxalase I (from E. coli)
Supplier: BioVision
AB168004-100UG
10008-500EA 827.42 USD InStock
10008-500
Human Recombinant Glyoxalase I (from E. coli)
Enzymes
>90% Pure Active Recombinant Human Glyoxalase I
Specific activity: >0.4 units/mg.
GLO1 is involved in the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. GLO1 is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere. GLO1 enzyme is abundantly expressed and present in numerous tumor cell lines, in which its concentration is often upregulated ubiquitously. GLO1 is a major susceptible gene for autism in an ethnic Chinese population from Taiwan. GLO1 might be involved in the pathophysiology of mood disorders. GLO1 plays a role in the pathophysiology of mood disorders. Overexpression of GLO1 is associated with kidney tumor.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty