Would you like to log in again?
Human Recombinant Scd100 (from CHO Cells)
Supplier: BioVision
AB285764-50UG AB285764-10UG
10009-164EA 528.59 USD InStock
10009-164 10009-166
Human Recombinant Scd100 (from CHO Cells)
Proteins and Peptides
>97% Pure active Human Recombinant sCD100.
Measured by its ability to inhibit chemokine (hMCP-3) induced human monocyte migration.
The Semaphorins are a large family of phylogenetically conserved proteins that play a pivotal role in maintaining homeostasis in the immune system. Twenty members of this family have been identified and categorized into eight subclasses based on sequence similarity and distinctive structural features. CD100, also known as Sema4D, is a 150 kDa transmembrane class IV semaphorin. Studies have shown that CD100 can induce monocyte migration, T-cell activation, and B-cell survival, as well as T/B cell and T/DC “cooperation”. The CD100 precursor contains 862 amino acids, including a 21 aa. signal sequence, a 713 aa. extracellular domain, a 21 aa transmembrane sequence, and a 107 aa cytoplasmic region. The extracellular sequence contains several structural features, including a 479 aa “sema” domain, a 79 aa. Ig-like sequence, and a 52 aa “Plexin-type repeat”. Recombinant soluble CD100 is a 78.9 kDa protein comprising the extracellular domain of CD100 (711 amino acids). SDS-PAGE analysis run under non-reducing conditions shows a mixture of disulfide linked dimer and monomer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty