Would you like to log in again?
Mouse Recombinant Lbp Mouse (from CHO Cells)
Supplier: BioVision
AB119732-10UG
10007-330EA 980.59 USD InStock
10007-330
Mouse Recombinant Lbp Mouse (from CHO Cells)
Proteins and Peptides
>95% Pure recombinant murine LBP
Up to 2 µg/ ml LBP mediates binding of FITC-LPS (0.5µg/ml) to CD14+CHO transfectants (2 x 106/ml)
Natural LBP is a 58KD glycoprotein produced in liver. It binds at lipid A of LPS with high affinity (10-9M) and reduced the cellular LPS effects at CD14+ cells (IL1ß, IL6, TNFα). It acts as opsonin for GRAM negative cells, LPS, neutrophiles and granulocytes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty