Would you like to log in again?
Human Recombinant Ubiquitin Activ Enzyme E1
Supplier: BioVision
AB285724-50UG
10008-526EA 1041.88 USD InStock
10008-526
Human Recombinant Ubiquitin Activ Enzyme E1
Proteins and Peptides
>95% Pure Human recombinant Ubiquitin Activating Enzyme E1
Typical enzyme concentration of 5 nM – 100 nM is used for in vitro conjugation, depending on conditions.
UBE1, the ubiquitin activating enzyme, is the first enzyme utilized in the pathway to conjugate ubiquitin onto target substrates through an ubiquitin-protein isopeptide bond. The C-terminal glycine of an ubiquitin (G76) is initially adenylated in an ATP dependent manner activating it so it can be transferred to the active site cysteine of UBE1 forming an ubiquitin-UBE1 thiolester. With the help of the other enzymes in the ubiquitin conjugation pathway (E2/E3 enzymes) the ubiquitin is transferred to a specific lysine on a target substrate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty