Would you like to log in again?
DeBakey Atraumatic Forceps, Full Curve, OR Grade, Sklar
Supplier: Sklar
52-6602
10196-478EA 666.74 USD InStock
10196-478
DeBakey Atraumatic Forceps, Full Curve, OR Grade, Sklar
Forceps
Sklar's DeBakey Atraumatic Forceps may be used as a hemostat to prevent blood flow, or as grasping forceps since the atraumatic teeth provide a secure grip but do not harm the tissues
This device has a full curve with 1x2 rows of fine, atraumatic teeth.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty