Would you like to log in again?
Cooley Atraumatic Neonatal Vascular Clamp, OR Grade, Sklar
Supplier: Sklar
52-6985
10197-104EA 777.51 USD InStock
10197-104
Cooley Atraumatic Neonatal Vascular Clamp, OR Grade, Sklar
Forceps
Sklar's Cooley Atraumatic Neonatal Vascular Clamp is a ratcheted, finger ring clamp designed for clamping vessels in newborns
The Cooley style atraumatic teeth cause little to no damage to the delicate vessels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty