Would you like to log in again?
Glutathione Reductase Activity Assay Kit, BioVision
Supplier: BioVision
AB83461-200
10013-790EA 1005.7 USD InStock
10013-790
Glutathione Reductase Activity Assay Kit, BioVision
Assays Enzyme Assays
Highly Sensitive Assay, HTS
  • Absorbance (405 nm)
  • Mammalian
  • Can detect 0.1 – 40 mU/ml GR in various samples
Glutathione Reductase (GR, EC 1.8.1.7) catalyzes the NADPH-dependent reduction of oxidized glutathione (GSSG) to reduced glutathione (GSH), which plays an important role in the GSH redox cycle that maintains adequate levels of reduced GSH. A high GSH/GSSG ratio is essential for protection against oxidative stress. BioVision’s Glutathione Reductase Assay Kit is a highly sensitive, simple, direct and HTS-ready colorimetric assay for measuring GR activity in biological samples. In the assay, GR reduces GSSG to GSH, which reacts with 5, 5′-Dithiobis (2-nitrobenzoic acid) (DTNB) to generate TNB2- (yellow color, λmax = 405 nm). The assay can detect 0.1 – 40 mU/ml GR in various samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty