Would you like to log in again?
Thioredoxin Reductase Activity Assay Kit, BioVision
Supplier: BioVision
AB83463-100
10013-794EA 531.35 USD InStock
10013-794
Thioredoxin Reductase Activity Assay Kit, BioVision
Assays Enzyme Assays
Thioredoxin reductase (TrxR) (EC 1.8.1.9) is a ubiquitous enzyme which is involved in many cellular processes such as cell growth, p53 activity, and protection against oxidation stress, etc.
  • Absorbance (412 nm)
  • Mammalian
The mammalian TrxR reduces thioredoxins as well as non-disulfide substrates such as selenite, lipoic acids, lipid hydroperoxides, and hydrogen peroxide. BioVision’s Thioredoxin Reductase Assay Kit provides a convenient colorimetric assay for detecting TrxR activity in various samples. In the assay TrxR catalyzes the reduction of 5, 5’-dithiobis (2-nitrobenzoic) acid (DTNB) with NADPH to 5-thio-2-nitrobenzoic acid (TNB2-), which generate a strong yellow color (λmax = 412 nm). Since in crude biological samples other enzymes, such as glutathione reductase and glutathione peroxidase, can also reduce DTNB, therefore, TrxR specific inhibitor is utilized to determine TrxR specific activity. Two assays are performed: the first measurement is of the total DTNB reduction by the sample, and the second one is the DTNB reduction by the sample in the presence of the TrxR specific inhibitor. The difference between the two results is the DTNB reduction by TrxR.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty