Would you like to log in again?
PAF Acetylhy Activity Assay Kit Colorimetric, BioVision
Supplier: BioVision
AB287879-96TEST
10013-796EA 643.52 USD InStock
10013-796
PAF Acetylhy Activity Assay Kit Colorimetric, BioVision
Assays Enzyme Assays
Highly Sensitive Assay
  • Absorbance (OD 412 nm)
  • Measurement of PAF-AH activity in various samples
  • Study/characterize PAF-AH inhibitors or activators
  • Mechanistic study of inflammatory disorders
Platelet-Activating Factor (PAF or 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) is an important phospholipid mediator, which has diverse biological activities. PAF is synthesized and secreted by a variety of cells such as mast cells, monocytes, macrophages etc. Upregulated PAF signaling can cause pathological inflammation and also has been found to be responsible for sepsis, shock, and traumatic injury. PAF Acetylhydrolase (PAF-AH or 1-alkyl-2-acetylglycerophosphocholine esterase or Lipoprotein-associated phospholipase A2 or Lp-PLA2) (EC 3.1.1.47) hydrolyzes PAF by removing acetyl group at the sn-2 position and converts PAF into biologically inactive form, lyso-PAF. PAF-AH has two forms: extracellular and intracellular that shares some similarities. In human, PAF-AH deficiency leads to severe asthma. Therefore early detection of PAF-AH activity is critical for mechanistic study, diagnosis, prevention, and therapeutic purpose. BioVision’s PAF Acetylhydrolase Assay kit provides a quick and easy way for monitoring PAF-AH activity in a variety of samples. In this kit, PAF-AH converts PAF into an inactivated form and probe gets converted into a colored product with strong absorbance at 412 nm. The assay is simple, sensitive, and high-throughput adaptable. Detection limit: < 0.1mU.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty