Would you like to log in again?
Protein Quantitation Kit, BioVision
Supplier: BioVision
AB102535-1000TEST
10013-802EA 641.45 USD InStock
10013-802
Protein Quantitation Kit, BioVision
Assays Protein Assays
The Protein Quantification Kit provides a simple and rapid procedure for determining the concentration of protein in solution.
  • Absorbance (595 nm)
  • Provides a simple and rapid procedure
  • Determine the concentration of protein in solution
The method utilizes an improved Coomassie blue G reagent which forms a blue complex in the presence of protein. The intensity of the blue complex is proportional to the amount of protein in the sample and can be easily measured by spectrophotometer or plate reader at 595 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty