Would you like to log in again?
Protein Carbonyl Content Assay Kit, BioVision
Supplier: BioVision
AB126287-100TEST
10013-808EA 778.95 USD InStock
10013-808
Protein Carbonyl Content Assay Kit, BioVision
Assays Enzyme Assays
Simple assay to quantify protein carbonyls
  • Absorbance ( 375 and 562 nm)
  • Mammalian
Protein carbonyl groups are an important and immediate biomarker of oxidative stress. DNPH tagging of protein carbonyls has been one of the most common measures of oxidative stress. DNP hydrazones formed from the reaction are easily quantifiable at 375 nm. BioVision’s Protein Carbonyl Content Assay Kit is designed to provide a simple and accurate method of quantifying carbonyls in protein samples. Using BSA as an example, a 1 mg (~ 15 nmol) sample has a detection limit of about 0.15 nmol carbonyl, where BSA typically contains approximately 1-3 nmol carbonyl/mg.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty