Would you like to log in again?
Laforce Foreign Body Spud, OR Grade, Sklar
Supplier: Sklar
65-7255
10193-384EA 160.24 USD InStock
10193-384
Laforce Foreign Body Spud, OR Grade, Sklar
Dissection Needles
Sklar's Laforce Foreign Body Spud is an ophthalmic instrument typically used for removing foreign objects and debris from the eye.
This instrument is often referred to as the "golf club" spud because the shaft and tip resemble a golf club. This product has a flat, straight handle that meets a thin, straight shaft that tapers to a slightly angled 2 mm tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty