Would you like to log in again?
Dissecting Spring Scissors, Noyes, Slight Curve Sharp, Roboz Surgical
Supplier: Roboz
RS-5675
100188-926EA 459.69 USD InStock
100188-926
Dissecting Spring Scissors, Noyes, Slight Curve Sharp, Roboz Surgical
Scissors
Noyes Micro Dissecting Spring Scissors; Slightly Curved, Sharp Points; 13 mm Cutting Edge; 0.25 mm Tip Width; 4 1/2" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty