Would you like to log in again?
Carbide Round Burrs Size 4, Hand Piece, Roboz Surgical
Supplier: Roboz
RS-6280C-4
100188-838BX 44.31 USD InStock
100188-838
Carbide Round Burrs Size 4, Hand Piece, Roboz Surgical
Dissection Sets
Carbide Round Burrs; Hand Piece; Size #4, 1.4 mm Tips; 1.75" Length; 2.33 mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill
These items are designed for animal research only and are not intended for human use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty