Would you like to log in again?
Micro Trephine, 2.30 mm, Tungsten Carbide, Roboz Surgical
Supplier: Roboz
RS-6308
100188-826EA 94.15 USD InStock
100188-826
Micro Trephine, 2.30 mm, Tungsten Carbide, Roboz Surgical
Dissection Sets
Micro Dissecting Trephine; Handpiece; Tungsten Carbide; 2.3 mm Diameter
The head diameter of these trephines is 2.3 mm. The shaft diameter is 2.33 mm. They work with our Ideal micro drill or other handpieces or drills accepting drill bits with 2.33 mm shaft diameter.
These items are designed for animal research only and are not intended for human use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty