Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deaver Scissors 5.5" Straight Sharp/Sharp, Roboz Surgical

Supplier: Roboz
RS-6762
100190-322EA 137.08 USD InStock
100190-322
Deaver Scissors 5.5" Straight Sharp/Sharp, Roboz Surgical
Scissors
Straight; Sharp-Sharp; 43 mm Blade Length; 5 1/2" Overall Length

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty