Would you like to log in again?
Dean Scissors 6.75" Angled Curved, Roboz Surgical
Supplier: Roboz
RS-7041
100190-242EA 218.77 USD InStock
100190-242
Dean Scissors 6.75" Angled Curved, Roboz Surgical
Scissors
Dean Scissors; Blades Angled On Flat (To The Right); One Serrated Blade; Slightly Curved Handles; 21 mm Blade Length; 6 3/4" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty