Would you like to log in again?
Garrigue Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3536 90-3537
10194-022EA 984.19 USD InStock
10194-022 10197-194
Garrigue Weighted Vaginal Speculum, OR Grade, Sklar
Specula
Sklar's Garrigue Weighted Vaginal Speculum is an OB/GYN instrument used for examination of, or procedures within, the vaginal canal or cervix.
  • Premium OR Grade
  • Reusable and latex-free
While the speculum is considered self-retaining due to the weight, and may allow the user's hands to be free, the blade is shallow and the weights available may not provide an adequate amount of retraction of the posterior wall. The practitioner can best determine if a weighted speculum is most appropriate for the exam or procedure to be completed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty