Would you like to log in again?
Zimmerman-Walton Expressor, OR Grade, Sklar
Supplier: Sklar
06-4192
10194-702EA 155.58 USD InStock
10194-702
Zimmerman-Walton Expressor, OR Grade, Sklar
Dissection Needles
Sklar's Zimmerman-Walton Expressor is a dermal instrument with a cupped end for expressing comedones and a tapered end that can hold a 30-gauge needle for lancing closed comedones.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty