Would you like to log in again?
Brantley-Turner Vaginal Retractor, OR Grade, Sklar
Supplier: Sklar
90-3470
10199-232EA 564.02 USD InStock
10199-232
Brantley-Turner Vaginal Retractor, OR Grade, Sklar
Retractors
Sklar's Brantley-Turner Vaginal Retractor is a handheld, lateral retractor typically used in conjunction with a traditional duckbill speculum in order to retract the uterine walls and provide the user with a better view of the cervix.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty