Would you like to log in again?
Heinlein Expressor, OR Grade, Sklar
Supplier: Sklar
06-4186
10194-944EA 79.63 USD InStock
10194-944
Heinlein Expressor, OR Grade, Sklar
Dissection Needles
This is a dermal instrument that features a lancet end for opening closed comedones and one spoon-shaped end with a hole for expressing comedones.
  • Reusable
  • Latex-free
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty