Would you like to log in again?
Crile Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-1160
10195-042EA 216.17 USD InStock
10195-042
Crile Needle Holder, OR Grade, Sklar
Forceps
Sklar's Crile Needle Holder is used to hold and guide intermediate weight needles during suturing
This product is straight with serrated tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty