Would you like to log in again?
Gillies Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-2662
10195-046EA 235.14 USD InStock
10195-046
Gillies Needle Holder, OR Grade, Sklar
Forceps
Gillies Needle Holder is designed for use with delicate suturing needles for closing wounds
Censitrac Ready products are industry standard and constructed of stainless steel.
This product has an offset design with curved, cross-serrated tips with scissors incorporated to make it a versatile tool, including plastic surgery procedures. The Gillies needle holder has on offset larger ring.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty