Would you like to log in again?
Ives Rectal Speculum, OR Grade, Sklar
Supplier: Sklar
80-2245
10196-334EA 382.84 USD InStock
10196-334
Ives Rectal Speculum, OR Grade, Sklar
Specula
Sklar® Ives Rectal Speculum, also known as the Ives-Fansler Anoscope, is used for examining the anal canal, hemorrhoidectomy, and rectal excision.
Non-latex, plastic obturator used to assist with insertion of the speculum. Diameter at the tip is 18 mm and diameter nearest the handle is 25 mm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty